EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Synthesis and biological evaluation of a novel series of 2,2-bisaminomethylated aurone analogues as anti-inflammatory and antimicrobial agents.
PMID 20430485
Abstract
This is the first report on aurones as a new class of drugs with anti-inflammatory and antimicrobial agents. A series of 2,2-bisaminomethylated aurone analogues (4a-j) were synthesized by Mannich reaction from 1,3,5-trimethoxybenzene in three steps. The structures of the newly synthesized compounds were confirmed by IR, (1)H NMR and mass spectral analysis. All the synthesized compounds were screened against the pro-inflammatory cytokines (TNF-alpha, IL-6) and antimicrobial (antibacterial and antifungal) activity. Compounds 4a, 4b, 4c, 4d, and 4e showed promising results against IL-6 at 10 microM concentration (74-100%). Compounds 4a, 4b and 4c were found to be active against TNF-alpha (76-100%) at 10 microM. Interestingly, all compounds have shown good antimicrobial activity. Compounds 4d, 4e and 4f showed excellent antimicrobial activity as compared with standard drugs.