EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Purification and characterization of a human liver arylacetamide deacetylase.
PMID 2043131
Abstract
Arylacetamide deacetylation is an important enzyme activity in the metabolic activation of arylamine substrates to ultimate carcinogens, best described as a carboxylesterase/amidase type of reaction. A 7-fold variation in the Vmax of 2-acetylaminofluorene deacetylation in 24 human livers was observed. An acetylaminofluorene deacetylase was purified 90 fold from human liver microsomes by PEG-fractionation, anion exchange and hydrophobic interaction chromatography. The purified 45kD protein showed no amino acid sequence homology to other carboxylesterases, neither in its N-terminus nor in tryptic peptides. Antibodies raised against the deacetylase recognized the protein with high specificity. This report thus describes the first arylacetamide deacetylase in human liver.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
62287 Lipase A Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~2 U/mg
L4777
Lipase acrylic resin, ≥5,000 U/g, recombinant, expressed in Aspergillus niger
62288 Lipase B Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~9 U/mg
62301 Lipase from Aspergillus niger, powder (fine), ~200 U/g
62285 Lipase from Aspergillus oryzae, lyophilized, powder, white, ~50 U/mg
75577 Lipase from Burkholderia sp., lyophilized powder, slightly beige, ~12 U/mg
02569 Lipase from Candida antarctica, lyophilized, powder, beige, ~0.3 U/mg
62302 Lipase from Candida rugosa, lyophilized, powder (fine), 15-25 U/mg
L1754 Lipase from Candida rugosa, Type VII, ≥700 unit/mg solid
62316 Lipase from Candida rugosa, powder, yellow-brown, ≥2 U/mg
L8525 Lipase from Candida rugosa, lyophilized powder, ≥40,000 units/mg protein
L3170
Lipase from Candida sp., recombinant, expressed in Aspergillus niger
L8906 Lipase from Mucor javanicus, lyophilized powder, ≥300 units/mg solid (using olive oil)
L9031 Lipase from Mucor miehei, lyophilized powder, ≥4,000 units/mg solid (using olive oil)
62298 Lipase from Mucor miehei, powder, slightly brown, ~1 U/mg
L0382 Lipase from porcine pancreas, Type VI-S, ≥20,000 units/mg protein, lyophilized powder
62309 Lipase from Pseudomonas cepacia, powder, light beige, ≥30 U/mg
28602 Lipase from Pseudomonas fluorescens, powder, slightly beige, ≥160 U/mg
L9518
Lipase from Pseudomonas sp., Type XIII, lyophilized powder, ≥15 units/mg solid
L4277 Lipase from Rhizomucor miehei, ≥20,000 U/g
62310 Lipase from Rhizopus niveus, powder (fine), ≥1.5 U/mg
L3001 Lipase from wheat germ, Type I, lyophilized powder, 5-15 units/mg protein
62306 Lipase from wheat germ, ~0.1 U/mg
73940 Lipase immobilized from Candida antarctica, beads, slightly brown, >2 U/mg