EMAIL THIS PAGE TO A FRIEND
Natural product research
Construction and expression of a dual vector for chemo-enzymatic synthesis of plant indole alkaloids in Escherichia coli.
PMID 20432158
Abstract
A dual vector (pQE-70-STR1-SG) containing coding regions of strictosidine synthase (STR1, EC 4.3.3.2) and strictosidine glucosidase (SG, EC 3.2.1.105) from the Indian medicinal plant Rauvolfia serpentina was constructed. Functional expression of the vector in Escherichia coli cells (M15 strain) was proven by isolation of prepurified enzyme extracts, which show both STR1 and SG activities. Incubation of the enzyme in the presence of tryptamine and secologanin delivered the indole alkaloid cathenamine, demonstrating functional co-expression of both STR1- and SG-cDNAs. Cathenamine reduction by sodium borohydride leading to tetrahydroalstonine revealed the chemo-enzymatic indole alkaloid synthesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
41111
Ajmalicine, ≥98.0% (HPLC)
C21H24N2O3