EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
Analytical applications of enzymatic growth of quantum dots.
PMID 20432413
Abstract
We have developed an analytical assay to detect the enzymatic activity of acetylcholine esterase and alkaline phosphatase based on the generation of quantum dots by enzymatic products. Acetylcholine esterase converts acetylthiocholine into thiocholine. The latter enhances the rate of decomposition of sodium thiosulfate into H(2)S, which in the presence of cadmium sulfate yields CdS quantum dots showing a time dependent exponential growth, typical of autocatalytic processes. This assay was also applied to detect acetylcholine esterase inhibitors. Alkaline phosphatase hydrolyzes thiophosphate and yields H(2)S, which instantly reacts with Cd(2+) to give CdS quantum dots. The formation of CdS quantum dots in both reactions was followed by fluorescence spectroscopy and showed dependence on the concentration of enzyme and substrate.