EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Rhodium(III)-catalyzed isoquinolone synthesis: the N-O bond as a handle for C-N bond formation and catalyst turnover.
PMID 20433170
Abstract
An external-oxidant-free process to access the isoquinolone motif via cross-coupling/cyclization of benzhydroxamic acid with alkynes is described. The reaction features a regioselective cleavage of a C-H bond on the benzhydroxamic acid coupling partner as well as a regioselective alkyne insertion. Mechanistic studies point out the important involvement of a N-O bond as a tool for C-N bond formation and catalyst turnover.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
412260
Benzhydroxamic acid, 99%
C7H7NO2