EMAIL THIS PAGE TO A FRIEND
Environmental pollution (Barking, Essex : 1987)
Determination of four fluoroquinolone antibiotics in tap water in Guangzhou and Macao.
PMID 20434248
Abstract
Four fluoroquinolone antibiotics (norfloxacin, ciprofloxacin, lomefloxacin, and enrofloxacin) in tap water in Guangzhou and Macao were analyzed using high performance liquid chromatography fluorescence detection. The results showed that all target antibiotics were detected in high rate both in Guangzhou (77.5%) and Macao (100%), ranging from 1.0 to 679.7 ng/L (SD
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L2906
Lomefloxacin hydrochloride
C17H19F2N3O3 · HCl