EMAIL THIS PAGE TO A FRIEND
Clinical therapeutics
Outpatient treatment of propionic acidemia-associated hyperammonemia with N-carbamoyl-L-glutamate in an infant.
PMID 20435240
Abstract
The aim of this research letter was to describe the use of N-carbamoyl-L-glutamate as first-line treatment of hyperammonemia in a 4-month-old female patient with propionic acidemia (PA). A 4-month-old female patient weighing 3.9 kg presented with decompensation with PA and initial hyperammonemia of 451 microg/dL. The patient was receiving a protein-restricted diet supplemented with L-carnitine at a dosage of 100 mg/kg/d during 2 consecutive months. Treatment with N-carbamoyl-L-glutamate was administered during 4 days on an outpatient basis at a dosage of 100 mg/kg/d, representing a dosage of 200 mg BID. The patient's hyperammonemia was monitored for 1 week. The patient's ammonia concentration started to decrease on the first day of the initiation of the treatment with N-carbamoyl-L-glutamate, from 451 to 68 microg/dL (normal range, 10-80 microg/dL) at day 6 of follow-up. The patient did not require intravenous fluids, invasive procedures, or hospitalization. N-carbamoyl-L-glutamate, combined with a protein-restricted diet and L-carnitine supplementation, was apparently effective as first-line treatment of hyperammonemia in this infant with PA.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C4375
N-Carbamyl-L-glutamic acid
C6H10N2O5