EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the USA
Electrophilic tuning of the chemoprotective natural product sulforaphane.
PMID 20439747
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
07-157
Anti-phospho-FAK (Tyr576) Antibody, serum, clone, Upstate®