EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
An improved route to PUGNAc and its galacto-configured congener.
PMID 20443616
Abstract
An efficient, scalable, and reliable synthesis of PUGNAc and its galacto-configured congener is reported.