EMAIL THIS PAGE TO A FRIEND
Zhong nan da xue xue bao. Yi xue ban = Journal of Central South University. Medical sciences
[Research progress in aldose reductase].
PMID 20448367
Abstract
Aldose reductase is a member of aldehyde-keto reductase superfamily widely existing in the kidney, adrenal gland, lens, retina, nerve, heart, placenta, brain, skeletal muscle, testis, blood vessels, lung, liver, et al. It is a reduced nicotinamide-adenine dinucleotide phosphate (NADPH)-dependent enzyme catalyzing the reduction of various aldehydes and ketones to the corresponding alcohol. It is involved in many oxidative stress diseases, cell signal transduction and cell proliferation process as well as diabetes complications. In recent years, some progress has been made in research of the activity and gene regulation of aldose reductase and the relation with many common diseases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0527 Epalrestat, ≥98% (HPLC)
C15H13NO3S2