EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Highly active gold-based catalyst for the reaction of benzaldehyde with ethyl diazoacetate.
PMID 20448977
Abstract
The gold complex [IPrAu(NCMe)]BF(4) catalyzes the reaction of ethyl diazoacetate with benzaldehyde to give mixtures of ethyl 3-oxo-3-phenylpropanoate and ethyl 3-hydroxy-2-phenylacrylate in the first example of a group 11 metal-based catalyst for this transformation; the catalyst activity is improved by a factor of 2500 compared to those of previously reported iron-based catalysts.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E22201
Ethyl diazoacetate, contains ≥13 wt. % dichloromethane
C4H6N2O2