EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Solvent properties of hydrazine in the preparation of metal chalcogenide bulk materials and films.
PMID 20449099
Abstract
A combination of unique solvent properties of hydrazine enables the direct dissolution of a range of metal chalcogenides at ambient temperature, rendering this an extraordinarily simple and soft synthetic approach to prepare new metal chalcogenide-based materials. The extended metal chalcogenide parent framework is broken up during this process, and the resulting metal chalcogenide building units are re-organized into network structures (from 0D to 3D) based upon their interactions with the hydrazine/hydrazinium moieties. This Perspective will review recent crystal and materials chemistry developments within this family of compounds and will briefly discuss the utility of this approach in metal chalcogenide thin-film deposition.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
751855
Hydrazine solution, 1 M in acetonitrile
H4N2