EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
A photosensing system composed of photosystem I, molecular wire, gold nanoparticle, and double surfactants in water.
PMID 20449307
Abstract
Photosensing performance of a system composed of photosystem I (PSI), vitamin K(1) (VK(1))-like molecular wire, and gold nanoparticles (AuNPs) in an aqueous solution was increased considerably by the addition of double surfactants, hexylamine and dodecylbenzenesulfonate.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
219703
Hexylamine, 99%
C6H15N