EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Analysis of aldehydes in beer by gas-diffusion microextraction: characterization by high-performance liquid chromatography-diode-array detection-atmospheric pressure chemical ionization-mass spectrometry.
PMID 20451914
Abstract
In this work, a recently developed extraction technique for sample preparation aiming the analysis of volatile and semi-volatile compounds named gas-diffusion microextraction (GDME) is applied in the chromatographic analysis of aldehydes in beer. Aldehydes-namely acetaldehyde (AA), methylpropanal (MA) and furfural (FA)-were simultaneously extracted and derivatized with 2,4-dinitrophenylhydrazine (DNPH), then the derivatives were separated and analyzed by high-performance liquid chromatography with spectrophotometric detection (HPLC-UV). The identity of the eluted compounds was confirmed by high-performance liquid chromatography-atmospheric pressure chemical ionization-mass-spectrometry detection in the negative ion mode (HPLC-APCI-MS). The developed methodology showed good repeatability (ca. 5%) and linearity as well as good limits of detection (AA-12.3, FA-1.5 and MA 5.4microgL(-1)) and quantification (AA-41, FA-4.9 and MA 18microgL(-1)); it also appears to be competitive in terms of speed and cost of analysis.