EMAIL THIS PAGE TO A FRIEND
Neurobiology of disease
A mGluR5 antagonist under clinical development improves L-DOPA-induced dyskinesia in parkinsonian rats and monkeys.
PMID 20452425
Abstract
L-DOPA remains the gold-standard treatment for Parkinson's disease but causes motor fluctuations and dyskinesia. Metabotropic glutamate receptor type 5 (mGluR5) has been proposed as a target for antidyskinetic therapies. Here, we evaluate the effects of fenobam, a noncompetitive mGluR5 antagonist already tested in humans, using rodent and nonhuman primate models of Parkinson's disease. In both animal models, acute administration of fenobam attenuated the L-DOPA-induced abnormal involuntary movements (50-70% reduction at the doses of 30mg/kg in rats and 10mg/kg in monkeys). The effect consisted in a reduction of peak-dose dyskinesia, whereas the end-dose phase was not affected. Chronic administration of fenobam to previously drug-naïve animals (de novo treatment) attenuated the development of peak-dose dyskinesia without compromising the anti-parkinsonian effect of L-DOPA. In addition, fenobam prolonged the motor stimulant effect of L-DOPA. We conclude that fenobam acts similarly in rat and primate models of L-DOPA-induced dyskinesia and represents a good candidate for antidyskinetic treatment in Parkinson's disease.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F0430
Fenobam, ≥98% (HPLC), solid
C11H11ClN4O2