EMAIL THIS PAGE TO A FRIEND
Archives of biochemistry and biophysics
Lower succinyl-CoA:3-ketoacid-CoA transferase (SCOT) and ATP citrate lyase in pancreatic islets of a rat model of type 2 diabetes: knockdown of SCOT inhibits insulin release in rat insulinoma cells.
PMID 20460097
Abstract
Succinyl-CoA:3-ketoacid-CoA transferase (SCOT) is a mitochondrial enzyme that catalyzes the reversible transfer of coenzyme-A from acetoacetyl-CoA to succinate to form acetoacetate and succinyl-CoA. mRNAs of SCOT and ATP citrate lyase were decreased 55% and 58% and enzyme activities were decreased >70% in pancreatic islets of the GK rat, a model of type 2 diabetes. INS-1 832/13 cells were transfected with shRNAs targeting SCOT mRNA to generate cell lines with reduced SCOT activity. Two cell lines with >70% knockdown of SCOT activity showed >70% reduction in glucose- or methyl succinate-plus-beta-hydroxybutyrate-stimulated insulin release. Less inhibition of insulin release was observed with two cell lines with less knockdown of SCOT. Previous studies showed knockdown of ATP citrate lyase in INS-1 832/13 cells does not lower insulin release. The results further support work that suggests mitochondrial pathways involving SCOT which supply acetoacetate for export to the cytosol are important for insulin secretion.