EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Transaminations with isopropyl amine: equilibrium displacement with yeast alcohol dehydrogenase coupled to in situ cofactor regeneration.
PMID 20461279
Abstract
Enantiopure chiral amines synthesis using omega-transaminases is hindered by an unfavourable equilibrium, but when using isopropylamine as the amine donor the equilibrium can be completely displaced by using a specific dehydrogenase in situ for removal of formed acetone.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
471291
Isopropylamine, ≥99.5%
C3H9N