EMAIL THIS PAGE TO A FRIEND
Advanced materials (Deerfield Beach, Fla.)
Photocatalytic cleavage of self-assembled organic monolayers by UV-induced charge transfer from GaN substrates.
PMID 20461720
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
376213
Trimethoxy(octadecyl)silane, technical grade, 90%
C21H46O3Si