EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Urea-stabilized air-stable Pt nanoparticles for thin film deposition.
PMID 20461854
Abstract
The reduction of [Pt(COD)(CH3)2] with CO in the presence of hexadecylamine (HDA) and oleic acid (OlAc) leads to amine carbonylation and formation of an air-stable colloidal solution of N,N'-bis(hexadecyl)urea-stabilized Pt(0) nanoparticles. These air-stable colloidal solutions can be used to form thin films of Pt nanoparticles on a silicon substrate.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H7408
Hexadecylamine, technical grade, 90%
C16H35N