EMAIL THIS PAGE TO A FRIEND
Drug metabolism and disposition: the biological fate of chemicals
Formation pathways of gamma-butyrolactone from the furan ring of tegafur during its conversion to 5-fluorouracil.
PMID 20463005
Abstract
Tegafur (FT) is a 5-fluorouracil (5-FU) prodrug that has been clinically used for various cancer chemotherapies. The following metabolites of FT were identified in patients: 5-FU, fluoro-beta-alanine, and gamma-butyrolactone (GBL) and its acidic form, gamma-hydroxybutyrate (GHB). GBL/GHB, which is probably generated from the furan ring of FT, inhibits tumor cell angiogenesis, contributing to the antitumor effect of FT-based therapies. In the present study, we identified the metabolites formed from the furan ring of FT by CYP2A6 and thymidine phosphorylase (TPase) using 2,4-dinitrophenylhydrazine derivatization procedures and clarified the metabolic pathway of FT to GBL/GHB. Succinaldehyde (SA) and 4-hydroxybutanal (4-OH-BTL) were produced as the metabolites because of the cleavage of the furan ring of FT during its conversion to 5-FU in cDNA-expressed CYP2A6 and purified TPase, respectively; however, GBL/GHB was hardly detected in cDNA-expressed CYP2A6 and purified TPase. GBL/GHB was formed after human hepatic microsomes or cDNA-expressed CYP2A6 mixed with cytosol were incubated with FT. Furthermore, 4-OH-BTL was converted to GBL/GHB in the microsomes and cytosol. These results suggest that GBL/GHB is generated from FT through the formation of SA and 4-OH-BTL but not directly from FT. Furthermore, the amount of 5-FU and GBL/GHB formed in the hepatic S9 was markedly decreased in the presence of a CYP2A6 inhibitor, suggesting that GBL/GHB may be mainly generated through the CYP2A6-mediated formation of SA.