EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
High activity and selectivity of Ag/SiO2 catalyst for hydrogenation of dimethyl oxalate.
PMID 20464002
Abstract
Ag/SiO(2) prepared by a sol-gel process is highly effective for selective gas-phase hydrogenation of dimethyl oxalate to corresponding alcohols. The catalysts are of great potential as industrially viable and novel catalysts for the production of methyl glycolate and ethylene glycol.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
135623
Dimethyl oxalate, ReagentPlus®, 99%
C4H6O4