EMAIL THIS PAGE TO A FRIEND
Biodegradation
Isolation and characterization of fenamiphos degrading bacteria.
PMID 20464454
Abstract
The biological factors responsible for the microbial breakdown of the organophosphorus nematicide fenamiphos were investigated. Microorganisms responsible for the enhanced degradation of fenamiphos were isolated from soil that had a long application history of this nematicide. Bacteria proved to be the most important group of microbes responsible for the fenamiphos biodegradation process. Seventeen bacterial isolates utilized the pure active ingredient fenamiphos as a carbon source. Sixteen isolates rapidly degraded the active ingredient in Nemacur 5GR. Most of the fenamiphos degrading bacteria were Microbacterium species, although Sinorhizobium, Brevundimonas, Ralstonia and Cupriavidus were also identified. This array of gram positive and gram negative fenamiphos degrading bacteria appeared to be pesticide-specific, since cross-degradation toward fosthiazate, another organophosphorus pesticide used for nematode control, did not occur. It was established that the phylogenetical relationship among nematicide degrading bacteria is closer than that to non-degrading isolates.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45483
Fenamiphos, PESTANAL®, analytical standard
C13H22NO3PS