EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Concise formal synthesis of (-)-salinosporamide A (marizomib) using a regio- and stereoselective epoxidation and reductive oxirane ring-opening strategy.
PMID 20465296
Abstract
Expedient access to a highly functionalized 2-pyrrolidinone (8), the gamma-lactam core of 20S proteasome inhibitor (-)-salinosporamide A (marizomib; NPI-0052; 1), using a regio- and stereoselective epoxide formation/reductive oxirane ring-opening strategy is presented. Notably, the sequential construction of the C-4, C-3, and C-2 stereocenters of 1 in a completely stereocontrolled fashion is a key feature of streamlining the synthesis of intermediate 12. A related strategy is also discussed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
83300
2-Pyrrolidinone, purum, ≥98.0% (GC)
C4H7NO