EMAIL THIS PAGE TO A FRIEND
Ultrasonics sonochemistry
Ultrasonic activation of Heck type reactions in the presence of Aliquat-336.
PMID 20471902
Abstract
Without phosphine and base, palladium catalysts such as PdCl(2), Pd(OAc)(2) and PdCl(2)(PhCN)(2) in water/DMF mixtures with Aliquat-336 proved to be excellent catalytic systems for Heck reactions involving several aryl bromides with styrene and acrylic compounds. Yields are remarkably improved under ultrasonic irradiation and the products show a high purity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
69485
Methyltrioctylammonium chloride, ≥97.0% (AT)
C25H54ClN