EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Pentenoic acid pathways for cellulosic biofuels.
PMID 20480484
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
245925
4-Pentenoic acid, 97%
C5H8O2
W284300
4-Pentenoic acid, ≥98%, stabilized, FG
C5H8O2