EMAIL THIS PAGE TO A FRIEND
Nano letters
Probing protein kinase (CK2) and alkaline phosphatase with CdSe/ZnS quantum dots.
PMID 20481436
Abstract
Semiconductor quantum dots (QDs) are used for the optical analysis of casein kinase (CK2) or the hydrolytic activity of alkaline phosphatase (ALP). Two schemes for the analysis of CK2 by a FRET-based mechanism are described. One approach involves the CK2-catalyzed phosphorylation of a serine-containing peptide (1), linked to CdSe/ZnS QDs, with Atto-590-functionalized ATP. The second analytical method involves the specific association of the Atto-590-functionalized antibody to the phosphorylated product. The hydrolytic activity of ALP is followed by the application of phosphotyrosine (4)-modified CdSe/ZnS QDs in the presence of tyrosinase as a secondary reporter biocatalyst. The hydrolysis of (4) yields the tyrosine units that are oxidized by O(2)/tyrosinase to the respective dopaquinone product. The latter quinone units quench the QDs via an electron transfer route, leading to the optical detection of the ALP activity.