EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Identification and functional characterization of a stable, centrally active derivative of the neurotensin (8-13) fragment as a potential first-in-class analgesic.
PMID 20481538
Abstract
The neurotensin hexapapetide fragment NT(8-13) is a potent analgesic when administered directly to the central nervous system but does not cross the blood-brain barrier. A total of 43 novel derivatives of NT(8-13) were evaluated, with one, ABS212 (1), being most active in four rat models of pain when administered peripherally. Compound 1 binds to human neurotensin receptors 1 and 2 with IC(50) of 10.6 and 54.2 nM, respectively, and tolerance to the compound in a rat pain model did not develop after 12 days of daily administration. When it was administered peripherally, serum levels and neurotensin receptor binding potency of 1 peaked within 5 min and returned to baseline within 90-120 min; however, analgesic activity remained near maximum for >240 min. This could be due to its metabolism into an active fragment; however, all 4- and 5-mer hydrolysis products were inactive. This pharmacokinetic/pharmacodynamic dichotomy is discussed. Compound 1 is a candidate for development as a first-in-class analgesic.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N5266
Neurotensin Fragment 8-13 acetate salt, ≥97% (HPLC)
C38H64N12O8