EMAIL THIS PAGE TO A FRIEND
Environmental science & technology
Influence of rhenium speciation on the stability and activity of Re/Pd bimetal catalysts used for perchlorate reduction.
PMID 20481620
Abstract
Recent work demonstrates reduction of aqueous perchlorate by hydrogen at ambient temperatures and pressures using a novel rhenium-palladium bimetal catalyst immobilized on activated carbon (Re/Pd-AC). This study examines the influence of Re speciation on catalyst activity and stability. Rates of perchlorate reduction are linearly dependent on Re content from 0-6 wt %, but no further increases are observed at higher Re contents. Surface-immobilized Re shows varying stability and speciation both in oxic versus H(2)-reducing environments and as a function of Re content. In oxic solutions, Re immobilization is dictated by sorption of the Re(VII) precursor, perrhenate (ReO(4)(-)), to activated carbon via electrostatic interactions. Under H(2)-reducing conditions, Re immobilization is significantly improved and leaching is minimized by ReO(4)(-) reduction to more reduced species on the catalyst surface. X-ray photoelectron spectroscopy shows two different Re binding energy states under H(2)-reducing conditions that correspond most closely to Re(V)/Re(IV) and Re(I) reference standards, respectively. The distribution of the two redox states varies with Re content, with the latter predominating at lower Re contents where catalyst activity is more strongly dependent on Re content. Results demonstrate that both lower Re contents and the maintenance of H(2)-reducing conditions are key elements in stabilizing the active Re surface species that are needed for sustained catalytic perchlorate treatment.