EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
Anharmonic vibrational analysis of 3,4-diaminopyridine and 3-aminopyridine by density functional theory calculations.
PMID 20483656
Abstract
In this work, we will report a combined experimental and theoretical study on molecular structure and vibrational analysis of 3,4-diaminopyridine (3,4-DAP) and 3-aminopyridine (3-AP). The Fourier transform infrared and Fourier transform Raman spectra of 3,4-DAP were recorded in the solid phase. The molecular geometry, harmonic vibrational wavenumbers of 3-AP and 3,4-DAP in the ground-state have been calculated by using MP2 and density functional methods (B3LYP) using 6-311++G(d,p) as basis set. Predicted electronic absorption spectra 3,4-DAP from TD-DFT calculation have been analyzed comparing with the experimental UV-vis spectrum. The calculated HOMO and LUMO energies show that charge transfer occur in the molecule. A detailed interpretation of the infrared spectra of 3-AP and 3,4-DAP is reported. The theoretical spectrograms for FTIR and FT-Raman spectra of the title molecules have also been constructed. Comparison of the experimental spectra with anharmonic vibrational wavenumbers indicates that B3LYP results are more accurate.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D7148
3,4-Diaminopyridine, ≥98%
C5H7N3
A78209
3-Aminopyridine, 99%
C5H6N2