EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Sodium bicarbonate induces crystalline wax generation, activates host-resistance, and increases imazalil level in rind wounds of oranges, improving the control of green mold during storage.
PMID 20486661
Abstract
Imazalil (IMZ) was quantified in the flavedo and albedo (Citrus fruits outer and inner tissue of the exocarp) of wounded and unwounded Valencia L. Olinda oranges following a 2 min immersion at 25 degrees C in 50, 100, or 250 microg mL(-1) of the fungicide mixture with or without 3% sodium bicarbonate (SBC). The addition of SBC significantly reduced the decay incidence throughout 30 d of storage at 10 degrees C with 95% RH and 6 d of simulated marketing period at 25 degrees C and 75% RH. In unwounded oranges, IMZ uptake was not changed by the coapplication of SBC, and the fungicide was predominantly recovered in the flavedo. To the contrary, in the albedo of wounded fruit, the residue level increased by about 6-fold when the fungicide was applied with SBC. When SBC was coapplied to wounded fruit, the phytoalexin scoparone was induced in the albedo and the accumulation was not affected by IMZ. When fruit was treated with SBC, scanning electron microscopy observations evidenced a production of crystalline wax patches with branched stripes and the magnitude was positively correlated to the salt concentration in the mixture. The generation as fast as 24 h post-treatment, and the different morphology of the new wax suggests a displacement of intracuticular waxes which can affect the fungicide sorption and diffusion coefficient into the rind.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32007
Imazalil, PESTANAL®, analytical standard
C14H14Cl2N2O
33997
Imazalil sulfate, PESTANAL®, analytical standard
C14H14Cl2N2O · H2SO4