EMAIL THIS PAGE TO A FRIEND
Journal of food science
Physical properties and oxidation rates of unrefined menhaden oil (Brevoortia patronus).
PMID 20492290
Abstract
Unrefined menhaden oil was evaluated for thermal and rheological properties and its temperature-dependent viscosity and lipid oxidation rate were determined. Peroxide value, free fatty acids, density, specific gravity, water activity, moisture content, and enthalpy of the unrefined menhaden oil were 5.70 meq/kg, 3.80%, 0.93 g/mL, 0.93%, 0.52%, 0.15%, and 20.2 kJ/kg, respectively. The melting point range of unrefined menhaden oil was found to be -69.5 to 27.21 degrees C. The menhaden oil exhibited non-Newtonian fluid behavior at lower temperatures (5 to 25 degrees C), while it behaved like a Newtonian fluid at 30 degrees C. The oil apparent viscosity at 5 degrees C (0.22 Pa.s) was significantly higher (P < 0.05) than that at 30 degrees C (0.033 Pa.s). The average magnitude of activation energy for viscosity of the unrefined menhaden oil was 50.37 kJ/mol. The predicted apparent viscosity agreed (R(2)= 0.9837) satisfactorily with the experimental apparent viscosity. The minimal lipid oxidation rate of the oil was found at 25 and 35 degrees C for 6 h, higher lipid oxidation rates were observed when the oil was heated for 6 h at 45 to 85 degrees C. The rate of lipid oxidation for unrefined menhaden oil was temperature dependent (R(2)= 0.9425). This study showed that the magnitude of the apparent viscosity and oxidation rate of the unrefined menhaden oil was greatly influenced by temperature.