EMAIL THIS PAGE TO A FRIEND
Journal of colloid and interface science
X-ray grazing incidence diffraction and Langmuir monolayer studies of the interaction of beta-cyclodextrin with model lipid membranes.
PMID 20493495
Abstract
The interactions of beta-CD with one component monolayers of cholesterol (chol), 1-stearoyl-sn-glycero-3-phosphocholine (lyso-PC), 1,2-dipalmitpyl-sn-phosphocholine (DPPC), sphingomyelin (SM) and the SM/chol and DPPC/chol mixtures have been investigated by the Langmuir monolayer technique and the synchrotron grazing incidence X-ray diffraction (GIXD). The investigated lipid monolayers have been studied with and without the 10(-3) M solution of beta-CD in the aqueous subphase. The surface pressure-area (pi-A) isotherms and the relaxation of the monolayers (surface pressure-time curves) were monitored. Our experiments reveal that there is not impact of beta-CD on the packing properties of the DPPC monolayers, while the presence of beta-CD in subphase changes the in-plane organization of SM molecules. Monolayers composed of pure chol molecules have been rapidly affected by the presence of the beta-CD in the subphase. Our data show that beta-CD can complex and desorb one-chain phospholipid (lyso-PC) but this process is relatively slow and, as indicated by the GIXD data, beta-CD molecules are present at the air/water interface. Subtraction of cholesterol by the beta-CD from mixed binary systems containing SM/chol (70/30, 50/50 and 30/70 mol ratio) and DPPC/chol (70/30 and 50/50 mol ratio) has also been investigated. Our experiments proved that cholesterol can be removed from the mixed monolayers only when it is unbound. The beta-CD was not capable to distract the monolayers of the SM/chol, forming a stable complex of the 2:1 stoichiometry (as observed in the model lipid raft). Interestingly, at the surface pressure of 30 mN/m also at the molar proportion of 50/50 no cholesterol removal was observed. This was interpreted by relatively strong SM/chol interactions and the tight packing of the mixed monolayer. For model membranes, in which cholesterol was in large excess (SM/chol, 30/70) the beta-CD extraction of cholesterol was observed, and the membrane composition evolves towards the lipid proportion corresponding to the stable complex stoichiometry (SM/chol 2:1).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L2131
1-Stearoyl-sn-glycero-3-phosphocholine, ≥99%, powder
C26H54NO7P