EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Discovery of a series of indan carboxylic acid glycogen phosphorylase inhibitors.
PMID 20493691
Abstract
A series of carboxylic acid glycogen phosphorylase inhibitors, which have potential as oral antidiabetic agents, is described. Defining and applying simple physicochemical design criteria was used to assess the opportunity and to focus synthetic efforts on compounds with the greatest probability of success. The study led to compound 17, which exhibits a good balance of properties including potent inhibition of recombinant human liver glycogen phosphorylase in vitro, a good DMPK profile including excellent bioavailability and low clearance and good in vivo activity in a glucagon challenge model of diabetes in Zucker rats.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45789
Indan, analytical standard
C9H10
I1804
Indan, 95%
C9H10