EMAIL THIS PAGE TO A FRIEND
Biotechnology letters
Accumulation of zirconium phosphate by a Serratia sp.: a benign system for the removal of radionuclides from aqueous flows.
PMID 20495949
Abstract
Metal phosphate deposited enzymatically on Serratia sp. has been used successfully for the removal of radionuclides from aqueous flows. Previous studies using biogenic hydrogen uranyl phosphate (HUP) on Serratia sp. biofilm showed removal of 100% of (90)Sr, (137)Cs, and (60)Co via their intercalation into biogenic HUP crystals. Zirconium phosphates (ZrP) offer a potential non-toxic and non-radioactive alternative to HUP for water decontamination. A method was developed for biomanufacturing ZrP. Biogenic ZrP removed ca. 100% of Sr(2+) and Co(2+) (0.5 mM) from solutions to a molar ratio at saturation of ca. 1:0.6 for both Zr:Sr and Zr:Co. The potential for drinking water decontamination via bio-ZrP is discussed with respect to bio-HUP and also other commercially available materials.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
464236
Zirconium(IV) hydrogenphosphate
H2O8P2Zr