EMAIL THIS PAGE TO A FRIEND
Organic letters
Synthesis of (-)-paroxetine via enantioselective phase-transfer catalytic monoalkylation of malonamide ester.
PMID 20499863
Abstract
A new enantioselective synthetic method of (-)-paroxetine is reported. (-)-Paroxetine could be obtained in 15 steps (95% ee and 9.1% overall yield) from N,N-bis(p-methoxyphenyl)malonamide tert-butyl ester via the enantioselective phase-transfer catalytic alkylation and the diastereoselective Michael addition as the key steps.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
129593
Malonamide, 97%
C3H6N2O2