EMAIL THIS PAGE TO A FRIEND
Journal of paediatrics and child health
Bilateral subdural hygromas and cephalhaematomas in male twins with severe myotubular myopathy caused by a Novel c.431delT (p.Leu144fs) mutation in MTM1 gene.
PMID 20500434