EMAIL THIS PAGE TO A FRIEND
Clinical cancer research : an official journal of the American Association for Cancer Research
Augmentation of radiation response by motesanib, a multikinase inhibitor that targets vascular endothelial growth factor receptors.
PMID 20507929