EMAIL THIS PAGE TO A FRIEND
The Consultant pharmacist : the journal of the American Society of Consultant Pharmacists
Health trends.
PMID 20511173
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0070
Salsalate, ≥98% (HPLC)
C14H10O5