EMAIL THIS PAGE TO A FRIEND
Nucleic Acids Research
Three prime exonuclease I (TREX1) is Fos/AP-1 regulated by genotoxic stress and protects against ultraviolet light and benzo(a)pyrene-induced DNA damage.
PMID 20511593