EMAIL THIS PAGE TO A FRIEND
Journal of oleo science
Supported liquid membrane transport of alkali metal cations by monoazacryptand with a partially fluorinated sidearm and the corresponding monoazacrown ethers.
PMID 20513970
Abstract
Monoazacryptand [20.18.18] (1), monoaza-15-crown-5 (2a), and monoaza-18-crown-6 (2b) with a partially fluorinated sidearm were newly prepared and their transport abilities were estimated in a supported liquid membrane containing a mixture of 2- (perfluorohexyl) ethyl alcohol and 2- (perfluorooctyl) ethyl alcohol. In competitive passive transport of K+, Na+, and Li+ under neutral conditions, the K+ selectivity increased in the order: ionophore 2a
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
188832
15-Crown-5, 98%
C10H20O5