EMAIL THIS PAGE TO A FRIEND
Indian journal of biochemistry & biophysics
Propoxur-induced acetylcholine esterase inhibition and impairment of cognitive function: attenuation by Withania somnifera.
PMID 20521626
Abstract
Propoxur (2-isopropoxyphenyl N-methylcarbamate) is widely used as an acaricide in agriculture and public health programs. Studies have shown that sub-chronic exposure to propoxur can cause oxidative stress and immuno-suppression in rats. Carbamates are also known to exhibit inhibitory effect on cholinesterase activity, which is directly related to their cholinergic effects. In the present study, the effect of Withania somnifera (Ashwagandha), a widely used herbal drug possessing anti-stress and immunomodulatory properties was studied on propoxur-induced acetylcholine esterase inhibition and impairment of cognitive function in rats. Male Wistar rats were divided into four groups. Group I was treated with olive oil and served as control. Group II was administered orally with propoxur (10 mg/kg b.wt.) in olive oil, group III received a combination of propoxur (10 mg/kg b.wt.) and W. somnifera (100 mg/kg b.wt.) suspension and group IV W. somnifera (100 mg/kg b.wt.) only. All animals were treated for 30 days. Cognitive behaviour was assessed by transfer latency using elevated plus maze. Blood and brain acetylcholine esterase (AChE) activity was also assessed. Oral administration of propoxur (10 mg/kg b.wt.) resulted in a significant reduction of brain and blood AChE activity. A significant prolongation of the acquisition as well as retention transfer latency was observed in propoxur-treated rats. Oral treatment of W. somnifera exerts protective effect and attenuates AChE inhibition and cognitive impairment caused by sub-chronic exposure to propoxur.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45644
Propoxur, PESTANAL®, analytical standard
C11H15NO3