EMAIL THIS PAGE TO A FRIEND
Anaesthesia
A comparison of the effect on QT interval between thiamylal and propofol during anaesthetic induction*.
PMID 20528837
Abstract
The aim of this study was to determine the effect of thiamylal and propofol on heart rate-corrected QT (QTc) interval during anaesthetic induction. We studied 50 patients undergoing lumbar spine surgery. Patients were administered 3 microgxkg(-1) fentanyl and were randomly allocated to receive 5 mgxkg(-1) thiamylal or 1.5 mgxkg(-1) propofol as an induction agent. Tracheal intubation was performed after vecuronium administration. Heart rate, mean arterial pressure, bispectral index score, and 12-lead electrocardiogram were recorded at the following time points: just before (T1) and 2 min after (T2) fentanyl administration; 2 min after anaesthetic administration (T3); 2.5 min after vecuronium injection (T4); and 2 min after intubation (T5). Thiamylal prolonged (p < 0.0001), but propofol shortened (p < 0.0001), the QTc interval.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T9799
Thiamylal, analytical standard
C12H18N2O2S