EMAIL THIS PAGE TO A FRIEND
Bioscience, biotechnology, and biochemistry
Characterization of a germination-accelerating factor from the silkworm (Bombyx mori Linnaeus) of entomopathogenic fungus Nomuraea rileyi (Farlow) Samson.
PMID 20530914
Abstract
The conidium of the entomopathogenic fungus, Nomuraea rileyi, has been found to germinate rapidly in the presence of a host insect-derived extract. This extract therefore appears to contain an important factor involved in host recognition by N. rileyi, although the substance (germination-accelerating factor, GAF) responsible for such unique germination behavior has yet to be identified. Our previous study was extended to the isolation of GAF from pupae of the silkworm, a host insect of N. rileyi. This present work subjects GAF to a structural analysis. The chemical structure of GAF is characterized as 2S-amino-tetradeca-4-ene-1,3R-diol (D-erythro-C(14)-sphingosine) based on spectroscopic data. An examination of the structure-activity relationship shows that the activity of D-erythro-C(14)-sphingosine was superior to that of sphingosines with shorter and longer carbon chains. It is suggested that the molecular species with a 14-carbon chain of a sphingosine is important for host recognition.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S7049
D-Sphingosine, synthetic
C18H37NO2