EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Inhibition of chorismate-utilising enzymes by 2-amino-4-carboxypyridine and 4-carboxypyridone and 5-carboxypyridone analogues.
PMID 20532401
Abstract
Several 2-amino-4-carboxypyridine, 4- and 5-carboxypyridone-based compounds were prepared and tested against three members of the chorismate-utilising enzyme family, anthranilate synthase, isochorismate synthase and salicylate synthase. Most compounds exhibited low micromolar inhibition of these three enzymes. The most potent inhibitor was a 4-carboxypyridone analogue bearing a lactate side chain on the pyridyl nitrogen which exhibited inhibition constants of 5, 91 and 54 muM against anthranilate synthase, isochorismate synthase and salicylate synthase respectively.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C1761
Chorismic acid from Enterobacter aerogenes, ≥80%
C10H10O6