EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Non-occupational allergic contact dermatitis from 2-N-octyl-4-isothiazolin-3-one in a Japanese mattress gel-sheet used for cooling.
PMID 20536482
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46078
2-Octyl-4-isothiazolin-3-one, PESTANAL®, analytical standard
C11H19NOS