EMAIL THIS PAGE TO A FRIEND
Letters in applied microbiology
Use of chemosensitization to overcome fludioxonil resistance in Penicillium expansum.
PMID 20536709
Abstract
To overcome fludioxonil resistance of Penicillium expansum, a mycotoxigenic fungal pathogen causing postharvest decay in apple, by using natural phenolic chemosensitizing agents. Fludioxonil-resistant mutants of P. expansum were co-treated with different oxidising and natural phenolic agents. Resistance was overcome by natural phenolic chemosensitizing agents targeting the oxidative stress-response pathway. These agents also augmented effectiveness of the fungicide, kresoxim-methyl. Results indicated that alkyl gallates target mitochondrial respiration and/or its antioxidation system. Fungal mitochondrial superoxide dismutase (Mn-SOD) plays a protective role against alkyl gallates. Natural chemosensitizing agents targeting the oxidative stress-response system, such as Mn-SOD, can synergize commercial fungicides. Redox-active compounds can serve as potent chemosensitizing agents to overcome resistance and lower effective dosages of fungicides. This can reduce costs with coincidental lowering of environmental and health risks.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46102
Fludioxonil, PESTANAL®, analytical standard
C12H6F2N2O2