EMAIL THIS PAGE TO A FRIEND
Journal of hazardous materials
Relevance of an electrochemical process prior to a biological treatment for the removal of an organophosphorous pesticide, phosmet.
PMID 20538412
Abstract
The aim of this study was to examine the feasibility of coupling an electrochemical pre-treatment with a biological step in order to degrade phosmet, an organophosphorous pesticide. Preliminary biodegradation experiments showed that the target molecule was not assimilated by activated sludge. The pre-treatment consisted of potentiostatic electrolysis (-1.3 V/SCE) in a flow cell. After only one pass (1 mL min(-1)), cyclic voltammetry with a vitreous carbon electrode showed a total phosmet reduction in neutral medium confirmed by thin layer chromatography, which also highlighted the presence of several by-products. H NMR spectra of the main by-product showed the absence of the aromatic ring, only the phosphorus part of phosmet has been identified and phosmet-oxon, a very toxic derivative, was not formed. Lower toxicity and higher biodegradability characterized the electrolyzed solution, EC(50) value increased from 7% to 58% and BOD(5) value increased from 4 to 9 mg O(2) L(-1) after electrolysis leading to an increase of the BOD(5) on COD ratio from 0.19 to 0.42 (limit of biodegradability, 0.4). These encouraging results were confirmed during activated sludge culture since an almost total mineralization of the electrolyzed solution was recorded (97%), confirming the feasibility of the proposed coupled process.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36195
Phosmet, PESTANAL®, analytical standard
C11H12NO4PS2