EMAIL THIS PAGE TO A FRIEND
Chemistry, an Asian journal
Local and remote charge-transfer-enhanced Raman scattering on one-dimensional transition-metal oxides.
PMID 20540066
Abstract
The one-dimensional (1D) transition-metal oxide MoO(3) belt is synthesized and characterized with X-ray diffraction, scanning electron microscopy, and Raman spectroscopy. Charge-transfer-(CT) enhanced Raman scattering of 4-mercaptobenzoic acid (4-MBA) on a 1D MoO(3) belt was investigated experimentally and theoretically. The chemical enhancement of surface-enhanced Raman scattering (SERS) of 4-MBA on the MoO(3) belt by CT is in the order of 10(3). The SERS of 4-MBA was investigated theoretically by using a quantum chemical method. The remote SERS of 4-MBA along the 1D MoO(3) belt (the light excitation to one side of the MoO(3) belt, and the SERS spectrum is collected on the other side of the MoO(3) belt) is also shown experimentally, which provides potential applications of SERS. The incident polarization dependence of remote SERS spectra has also been investigated experimentally.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
662534
4-Mercaptobenzoic acid, technical grade, 90%
C7H6O2S
706329
4-Mercaptobenzoic acid, 99%
C7H6O2S