EMAIL THIS PAGE TO A FRIEND
Advanced materials (Deerfield Beach, Fla.)
Flash nano-welding: investigation and control of the photothermal response of ultrathin bismuth sulfide nanowire films.
PMID 20540105
Abstract
Ultrathin Bi₂S₃ nanowires undergo a pronounced photothermal response to irradiation from a commercial camera flash. Controlled nano-welding was shown by using single walled carbon nanotube mats as an electrically and thermally conductive substrate. The resulting welded nanowire film is denser and has significantly lower resistance than unflashed bilayer films.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
333190
Bismuth(III) sulfide, 99%
Bi2S3