EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
A universal approach to the synthesis of noble metal nanodendrites and their catalytic properties.
PMID 20540128
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I7632
Iodobenzene, 98%
C6H5I